Assay Design



Gene Transcripts

Gene Symbol
MRP
Entrez Gene ID
34686
Gene Name
Multidrug-Resistance like Protein 1
Gene Aliases
ABCC1, CG6214, Dmel\CG6214, Dmel_CG6214, MRP1, Mrp, Mrp1, dMRP, dMRP/CG6214, dMRP1, dmrp, lincRNA.123
Chromosome Location
Chr.2L: 12719045 - 12766546 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032456
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_165020.3 NP_723772.2 8 - 9 3503 74
NM_205971.2 NP_995693.1 8 - 9 3499 74
NM_205980.2 NP_995702.1 9 - 10 3617 74

View Full Product Details