Assay Design



Gene Transcripts

Gene Symbol
yuri
Entrez Gene ID
34894
Gene Name
yuri gagarin
Gene Aliases
BG:DS05639.1, BG:DS07851.11, CG11860, CG12635, CG31732, Dmel\CG31732, Dmel_CG31732, Yuri
Chromosome Location
Chr.2L: 15257267 - 15264692 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0045842
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316397.1 NP_001303326.1 3 - 4 916 59
NM_165106.5 NP_723903.3 2 - 3 794 59
NM_165107.3 NP_723904.1 2 - 3 1259 59
NM_165109.3 NP_723906.1 3 - 4 1204 59
GenBank mRNA BT001564.1 - 2 - 3 1257 59

View Full Product Details