Assay Design



Gene Transcripts

Gene Symbol
Hcf
Entrez Gene ID
43788
Gene Name
Host cell factor
Gene Aliases
CG1710, Dmel\CG1710, Dmel_CG1710, HCF, HCF1, HCF_DROME, Hcf1, anon-WO0172774.48, dHCF, dHcf, dHcf1
Chromosome Location
Chr.4: 359938 - 375554 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039904
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258491.3 NP_001245420.1 12 - 13 4471 89
NM_079882.4 NP_524621.2 11 - 12 4445 89
NM_166756.3 NP_726566.1 11 - 12 4141 89
NM_166757.3 NP_726567.2 11 - 12 4289 89
NM_205873.3 NP_995595.1 11 - 12 4141 89
GenBank mRNA AJ320236.1 - 10 - 11 4098 89
BT015268.1 - 11 - 12 4406 89

View Full Product Details