Assay Design



Gene Transcripts

Gene Symbol
Eaat2
Entrez Gene ID
33247
Gene Name
Excitatory amino acid transporter 2
Gene Aliases
CG3159, Dmel\CG3159, Dmel_CG3159, EAAT, EAAT2, Eaat 2, dEAAT2, dEaat2
Chromosome Location
Chr.2L: 737212 - 744395 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026438
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169373.2 NP_001162844.1 2 - 3 1323 68
NM_001169374.2 NP_001162845.1 4 - 5 1094 68
NM_058079.3 NP_477427.1 4 - 5 982 68

View Full Product Details