Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001273036.1 | NP_001259965.1 | 8 - 9 | 2444 | 86 |
NM_001298638.1 | NP_001285567.1 | 11 - 12 | 1886 | 86 | |
NM_001298639.1 | NP_001285568.1 | 10 - 11 | 1883 | 86 | |
NM_001370000.1 | NP_001356906.1 | 11 - 12 | 2489 | 86 | |
NM_078736.3 | NP_523460.2 | 10 - 11 | 1889 | 86 | |
NM_164501.2 | NP_722838.1 | 10 - 11 | 1883 | 86 | |
NM_164502.2 | NP_722839.1 | 9 - 10 | 1532 | 86 |