Assay Design



Gene Transcripts

Gene Symbol
Aasdh
Entrez Gene ID
34175
Gene Name
Aminoadipate-semialdehyde dehydrogenase
Gene Aliases
CG13401, Dmel\CG13401, Dmel_CG13401, U26
Chromosome Location
Chr.2L: 8445861 - 8449576 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027780
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298825.1 NP_001285754.1 4 - 5 1484 77
NM_135386.5 NP_609230.2 4 - 5 1473 77
NM_205938.2 NP_995660.2 4 - 5 1473 77

View Full Product Details