Assay Design
Gene Transcripts
- Gene Symbol
- Rat1
- Entrez Gene ID
-
33964
- Gene Name
- Rat1 5'-3' exoribonuclease
- Gene Aliases
-
CG10354, Dmel\CG10354, Dmel_CG10354, RAT1, XRN2, Xrn2, dRAT1, dhp1
- Chromosome Location
-
Chr.2L: 6908083 - 6911423 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0031868
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135238.3 |
NP_609082.1 |
1 - 1 |
2930 |
150 |
GenBank mRNA |
BT056259.1 |
- |
1 - 1 |
2924 |
150 |
View Full Product Details