Assay Design
Gene Transcripts
- Gene Symbol
- PrBP
- Entrez Gene ID
-
34196
- Gene Name
- Prenyl-binding protein
- Gene Aliases
-
CG9296, DmPrBP, DmPrBP/delta, Dmel\CG9296, Dmel_CG9296
- Chromosome Location
-
Chr.2L: 8684823 - 8685955 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0032059
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135402.3 |
NP_609246.1 |
1 - 1 |
1043 |
122 |
GenBank mRNA |
AY071756.1 |
- |
1 - 1 |
1042 |
122 |
View Full Product Details