Assay Design



Gene Transcripts

Gene Symbol
nmd
Entrez Gene ID
44021
Gene Name
no mitochondrial derivative
Gene Aliases
CG5395, DmCG5395, Dmel\CG5395, Dmel_CG5395, Msp, Msp1, Nmd, anon-EST:Liang-1.79, clone 1.79, l(2)08774, ms(2)4, ms(2)ry4
Chromosome Location
Chr.2L: 10302173 - 10303947 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0005322
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298872.1 NP_001285801.1 1 - 2 297 108
NM_135529.3 NP_609373.1 1 - 2 297 108
GenBank mRNA AY119493.1 - 1 - 2 261 108

View Full Product Details