Assay Design



Gene Transcripts

Gene Symbol
AstCC
Entrez Gene ID
34538
Gene Name
Allatostatin double C
Gene Aliases
ASCC, ASTCC, Ast-CC, CG14920, Dmel\CG14920, Dmel_CG14920
Chromosome Location
Chr.2L: 11083459 - 11085229 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032337
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298901.1 NP_001285830.1 2 - 3 602 67
NM_135639.4 NP_609483.2 2 - 3 602 67

View Full Product Details