Assay Design



Gene Transcripts

Gene Symbol
tadr
Entrez Gene ID
35370
Gene Name
torn and diminished rhabdomeres
Gene Aliases
CG9264, Dmel\CG9264, Dmel_CG9264, torn
Chromosome Location
Chr.2L: 21075405 - 21081053 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032911
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_136238.3 NP_610082.2 6 - 7 1039 71
NM_165354.2 NP_724308.2 6 - 7 1039 71

View Full Product Details