Assay Design
Gene Transcripts
- Gene Symbol
- Gr39b
- Entrez Gene ID
-
117347
- Gene Name
- Gustatory receptor 39b
- Gene Aliases
-
39D.1, 39D1, CG31620, Dmel\CG31620, Dmel_CG31620, GR39D.1, GR39b, Gr39D1
- Chromosome Location
-
Chr.2L: 21367908 - 21369706 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0041245
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_165376.1 |
NP_724336.1 |
2 - 3 |
1055 |
105 |
View Full Product Details