Assay Design



Gene Transcripts

Gene Symbol
Df31
Entrez Gene ID
35418
Gene Name
Decondensation factor 31
Gene Aliases
BEST:LD04967, CG2207, DF 31, DF31, Dmel\CG2207, Dmel_CG2207, LD04967, anon-EST:Liang-1.46, anon-EST:Posey126, anon-EST:fe1A4, anon-WO0153538.53, anon1A4, clone 1.46, l(2)k05815
Chromosome Location
Chr.2L: 21626309 - 21629932 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0022893
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032217.3 NP_001027388.1 1 - 2 271 59
NM_078890.4 NP_523614.1 1 - 2 271 59
NM_165388.4 NP_724350.1 2 - 3 163 59
GenBank mRNA AF005846.1 - 1 - 2 396 59
AY051664.1 - 1 - 2 236 59

View Full Product Details