Assay Design



Gene Transcripts

Gene Symbol
scaf
Entrez Gene ID
35505
Gene Name
scarface
Gene Aliases
CG11066, CG15900, Dmel\CG11066, Dmel_CG11066, SPH142, Scaf, cSPH142, scarf
Chromosome Location
Chr.2R: 5676750 - 5691723 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033033
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_136336.3 NP_610180.1 2 - 3 431 70
NM_165441.2 NP_724424.1 2 - 3 778 70
GenBank mRNA AY047548.1 - 2 - 3 403 70

View Full Product Details