Assay Design



Gene Transcripts

Gene Symbol
Src42A
Entrez Gene ID
35524
Gene Name
Src oncogene at 42A
Gene Aliases
CG14471, CG44128, CG7873, DSrc42A, Dm SRC41, DmHD-29, Dmel\CG44128, Dmel_CG44128, Dsrc41, Dsrc42A, Dtk5, HD-29, SK2-4, SRC 42A, SRC42A, Src, Src41, Src42, Src42a, Su(D-raf)1, Su(Raf)1, Su(phl)1, Su1, Tk5, dSrc, dSrc42A, dtk-5, dtk5, l(2)k10108, src, src42, src42A, src42a
Chromosome Location
Chr.2R: 5981844 - 6012795 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264959
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057501.4 NP_476849.1 2 - 3 965 73
GenBank mRNA AY058652.1 - 2 - 3 929 73
BT044565.1 - 3 - 4 833 73

View Full Product Details