Assay Design



Gene Transcripts

Gene Symbol
esn
Entrez Gene ID
53557
Gene Name
espinas
Gene Aliases
CG12833, CG12835, CG43722, Dm_2R:11728, Dmel\CG43722, Dmel_CG43722, Esn
Chromosome Location
Chr.2R: 7086316 - 7127868 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0263934
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273831.1 NP_001260760.1 4 - 5 2050 108
NM_001273832.1 NP_001260761.1 5 - 6 1324 108
NM_080251.4 NP_524990.1 5 - 6 1634 108
NM_165507.2 NP_724533.1 5 - 6 1324 108
GenBank mRNA AJ251892.1 - 5 - 6 1637 108
BT030763.1 - 5 - 6 1324 108

View Full Product Details