Assay Design



Gene Transcripts

Gene Symbol
scra
Entrez Gene ID
35696
Gene Name
scraps
Gene Aliases
ABP8, Abp8, Ani, Anil, Anillin, CG2092, Dmel\CG2092, Dmel_CG2092, Scra, ani, anil, anillin, l(2)03427, l(2)43Ec, l(2)k08255
Chromosome Location
Chr.2R: 7590119 - 7595242 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261385
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_165543.2 NP_724582.1 6 - 7 2270 96
NM_165544.2 NP_724583.1 7 - 8 2106 96
GenBank mRNA AY069520.1 - 6 - 7 2126 96

View Full Product Details