Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144147.2 | NP_001137619.1 | 6 - 7 | 3487 | 88 |
NM_001273877.1 | NP_001260806.1 | 6 - 7 | 3487 | 88 | |
NM_136542.3 | NP_610386.2 | 5 - 6 | 2527 | 88 | |
NM_165609.3 | NP_724683.2 | 4 - 5 | 1581 | 88 |