Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001299285.1 | NP_001286214.1 | 10 - 11 | 3661 | 80 |
NM_001299286.1 | NP_001286215.1 | 10 - 11 | 3664 | 80 | |
NM_136570.4 | NP_610414.1 | 9 - 10 | 3604 | 80 | |
NM_165630.3 | NP_724716.1 | 9 - 10 | 3607 | 80 |