Assay Design
Gene Transcripts
- Gene Symbol
- Pgm2a
- Entrez Gene ID
-
35923
- Gene Name
- Phosphoglucomutase 2a
- Gene Aliases
-
CG8073, Dmel\CG8073, Dmel_CG8073, Pmm45A
- Chromosome Location
-
Chr.2R: 9107151 - 9109718 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033377
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136609.5 |
NP_610453.1 |
2 - 3 |
589 |
87 |
View Full Product Details