Assay Design



Gene Transcripts

Gene Symbol
hig
Entrez Gene ID
35949
Gene Name
hikaru genki
Gene Aliases
CG2040, CT6282, Dmel\CG2040, Dmel_CG2040, HIG
Chromosome Location
Chr.2R: 9227785 - 9247249 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010114
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014514.2 NP_001014514.1 4 - 5 1943 54
NM_165667.2 NP_724773.1 4 - 5 1943 54

View Full Product Details