Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057547.5 | NP_476895.1 | 4 - 5 | 683 | 87 |
NM_165694.3 | NP_724822.1 | 4 - 5 | 808 | 87 | |
NM_165695.2 | NP_724823.2 | 4 - 5 | 670 | 87 | |
GenBank mRNA | BT099607.1 | - | 4 - 5 | 654 | 87 |