Assay Design



Gene Transcripts

Gene Symbol
Cdc2rk
Entrez Gene ID
36051
Gene Name
Cdc2-related kinase
Gene Aliases
CG1362, Dcdrk, Dmel\CG1362, Dmel_CG1362, cdc2rk
Chromosome Location
Chr.2R: 10067217 - 10068702 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0013435
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078950.4 NP_523674.1 4 - 5 1095 77
GenBank mRNA BT089018.1 - 4 - 5 1052 77
BT099569.1 - 4 - 5 1087 77

View Full Product Details