Assay Design



Gene Transcripts

Gene Symbol
Pkd2
Entrez Gene ID
34651
Gene Name
Polycystic kidney disease 2
Gene Aliases
AMO, Amo, CG6504, Dmel\CG6504, Dmel_CG6504, Dmpkd2, PKD2, TRPP, TRPP2, amo, pkd2
Chromosome Location
Chr.2L: 12398274 - 12402023 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0041195
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_135717.3 NP_609561.2 8 - 9 2779 138
GenBank mRNA AY283154.1 - 8 - 9 2753 138
BT126366.1 - 8 - 9 2779 138

View Full Product Details