Assay Design



Gene Transcripts

Gene Symbol
dgo
Entrez Gene ID
36139
Gene Name
diego
Gene Aliases
CG12342, DGO, Dgo, Dmel\CG12342, Dmel_CG12342, EP(2)2619, EP2619
Chromosome Location
Chr.2R: 10816037 - 10819884 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0086898
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_136770.2 NP_610614.1 2 - 3 260 57
NM_165800.2 NP_724973.1 2 - 3 657 57
GenBank mRNA AY128436.1 - 2 - 3 223 57

View Full Product Details