Assay Design
Gene Transcripts
- Gene Symbol
- dy
- Entrez Gene ID
-
32130
- Gene Name
- dusky
- Gene Aliases
-
CG9355, Dmel\CG9355, Dmel_CG9355, ala, dy1
- Chromosome Location
-
Chr.X: 11769769 - 11778784 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004511
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078575.3 |
NP_511130.2 |
3 - 4 |
1260 |
64 |
GenBank mRNA |
BT016046.1 |
- |
3 - 4 |
1231 |
64 |
View Full Product Details