Assay Design



Gene Transcripts

Gene Symbol
LIMK1
Entrez Gene ID
32207
Gene Name
LIM domain kinase 1
Gene Aliases
CG1848, D-LIMK1, D-Limk, DLIMK, DLIMK1, Dlimk, Dmel\CG1848, Dmel_CG1848, LIM-kinase 1, LIMK, LIMK-1, LimK, LimK1, Limk, ag, agn, dLIMK, dLIMK1, l(1)11Af, limK1, limk, limk1, ts398
Chromosome Location
Chr.X: 12584385 - 12592193 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0283712
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169263.1 NP_001162734.1 1 - 2 93 94
NM_078584.3 NP_511139.2 3 - 4 973 94

View Full Product Details