Assay Design
Gene Transcripts
- Gene Symbol
- dmrt11E
- Entrez Gene ID
-
32291
- Gene Name
- doublesex-Mab related 11E
- Gene Aliases
-
CG15749, Dmel\CG15749, Dmel_CG15749, Dmrt11E, dmrt11
- Chromosome Location
-
Chr.X: 13368191 - 13370472 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0030477
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078591.3 |
NP_511146.2 |
2 - 3 |
814 |
84 |
View Full Product Details