Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258738.2 | NP_001245667.1 | 3 - 4 | 2029 | 66 |
NM_001298314.1 | NP_001285243.1 | 3 - 4 | 2123 | 66 | |
NM_078599.5 | NP_511154.3 | 3 - 4 | 2029 | 66 | |
GenBank mRNA | AY089580.1 | - | 3 - 4 | 2026 | 66 |