Assay Design



Gene Transcripts

Gene Symbol
NetA
Entrez Gene ID
32398
Gene Name
Netrin-A
Gene Aliases
CG18657, CT27014, Dmel\CG18657, Dmel_CG18657, NetrinA, net, netA, netrin, netrin A
Chromosome Location
Chr.X: 14604012 - 14653928 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015773
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298314.1 NP_001285243.1 4 - 5 2283 75
NM_078599.5 NP_511154.3 4 - 5 2189 75

View Full Product Details