Assay Design



Gene Transcripts

Gene Symbol
CycD
Entrez Gene ID
32551
Gene Name
Cyclin D
Gene Aliases
CDI3, CG9096, CYCD, Cdi3, Cyc D, Cyc-D, DmcycD, Dmel\CG9096, Dmel_CG9096, cdi3, cycD, cyclin D, cyclinD, dCycD
Chromosome Location
Chr.X: 15906822 - 15909676 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010315
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078631.4 NP_523355.2 2 - 3 837 61
NM_167472.3 NP_727912.2 2 - 3 948 61
NM_167473.2 NP_727913.2 1 - 2 1145 61

View Full Product Details