Assay Design
Gene Transcripts
- Gene Symbol
- betaCOP
- Entrez Gene ID
-
32820
- Gene Name
- Coat Protein (coatomer) beta
- Gene Aliases
-
BcDNA.GH09317, BcDNA:GH09317, Bcop, Beta Cop, BetaCop, CG6223, COPI, Dmel\CG6223, Dmel_CG6223, bcop, beta COP, beta-COP, beta-Cop, beta-CopII, betaCop
- Chromosome Location
-
Chr.X: 18381549 - 18385242 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0008635
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078676.4 |
NP_523400.1 |
4 - 5 |
2870 |
64 |
GenBank mRNA |
AF145656.1 |
- |
4 - 5 |
2823 |
64 |
View Full Product Details