Assay Design



Gene Transcripts

Gene Symbol
Sep1
Entrez Gene ID
33114
Gene Name
Septin 1
Gene Aliases
38960, CG1403, DSep1, Diff6, Dmel\CG1403, Dmel_CG1403, anon-19Fa, dSEPT1, iby, sep1, tu1
Chromosome Location
Chr.X: 21358286 - 21361688 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011710
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078706.4 NP_523430.1 4 - 5 920 99
GenBank mRNA AY121679.1 - 4 - 5 932 99
X67202.1 - - 943 99

View Full Product Details