Assay Design



Gene Transcripts

Gene Symbol
ds
Entrez Gene ID
33245
Gene Name
dachsous
Gene Aliases
CG17941, CT39575, DS, Dmel\CG17941, Dmel_CG17941, Ds, EP-822, cad2, dachs, l(2)01855, l(2)05142
Chromosome Location
Chr.2L: 640013 - 714983 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0284247
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298622.1 NP_001285551.1 8 - 9 6060 70
NM_078722.3 NP_523446.2 8 - 9 6060 70

View Full Product Details