Assay Design
Gene Transcripts
- Gene Symbol
- cup
- Entrez Gene ID
-
33934
- Gene Name
- CG11181-PB
- Gene Aliases
-
CG11181, CUP, Cup, DmCup, Dmel\CG11181, Dmel_CG11181, fs(1)cup, fs(2)cup, i239
- Chromosome Location
-
Chr.2L: 6663664 - 6674780 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000392
- Gene Symbol
- CG34310
- Entrez Gene ID
-
5740536
- Gene Name
- CG34310-PA
- Gene Aliases
-
Dmel\CG34310, Dmel_CG34310
- Chromosome Location
-
Chr.2L: 6663664 - 6674780 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0085339
View Full Product Details