Assay Design
Gene Transcripts
- Gene Symbol
- Ns4
- Entrez Gene ID
-
35338
- Gene Name
- Nucleostemin 4
- Gene Aliases
-
38E.14, CG9320, Dmel\CG9320, Dmel_CG9320, NS4, dmNS4, ns4
- Chromosome Location
-
Chr.2L: 20752476 - 20754655 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0032882
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136211.4 |
NP_610055.1 |
1 - 2 |
218 |
65 |
GenBank mRNA |
AY118506.1 |
- |
1 - 2 |
182 |
65 |
View Full Product Details