Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042910.3 | NP_001036375.2 | 4 - 5 | 1563 | 84 |
NM_001169570.2 | NP_001163041.1 | 3 - 4 | 1824 | 84 | |
NM_001273749.1 | NP_001260678.1 | 5 - 6 | 2292 | 84 | |
NM_136276.4 | NP_610120.1 | 3 - 4 | 515 | 84 | |
GenBank mRNA | AY071322.1 | - | 3 - 4 | 449 | 84 |
BT072836.1 | - | 3 - 4 | 467 | 84 | |
BT124891.1 | - | 5 - 6 | 2191 | 84 |