Assay Design



Gene Transcripts

Gene Symbol
dpr12
Entrez Gene ID
50320
Gene Name
defective proboscis extension response 12
Gene Aliases
CG14469, CG34385, CG7796, CT34165, Dmel\CG34385, Dmel_CG34385, Dpr-12, Dpr12
Chromosome Location
Chr.2R: 5804712 - 5860831 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0085414
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273787.1 NP_001260716.1 7 - 8 2338 58
NM_136342.2 NP_610186.2 6 - 7 2263 58
NM_144205.5 NP_652462.3 8 - 9 2384 58
GenBank mRNA BT029948.1 - 6 - 7 1514 58

View Full Product Details