Assay Design
Gene Transcripts
- Gene Symbol
- Spn43Ad
- Entrez Gene ID
-
35639
- Gene Name
- Serpin 43Ad
- Gene Aliases
-
CG1859, Dmel\CG1859, Dmel_CG1859
- Chromosome Location
-
Chr.2R: 7153738 - 7155424 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0044011
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136417.3 |
NP_610261.1 |
1 - 2 |
1027 |
66 |
GenBank mRNA |
AY071238.1 |
- |
1 - 2 |
1025 |
66 |
View Full Product Details