Assay Design



Gene Transcripts

Gene Symbol
eIF3b
Entrez Gene ID
36981
Gene Name
eukaryotic translation initiation factor 3 subunit b
Gene Aliases
CG4878, Dmel\CG4878, Dmel_CG4878, cg4878, eIF3 S9, eIF3-S9, eIF3-s9, eIF3B
Chromosome Location
Chr.2R: 17536213 - 17539142 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034237
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_137384.3 NP_611228.1 3 - 4 559 67
NM_166239.4 NP_725691.1 2 - 3 564 67
GenBank mRNA BT072816.1 - 2 - 3 684 67

View Full Product Details