Assay Design
Gene Transcripts
- Gene Symbol
- Rgk1
- Entrez Gene ID
-
14462605
- Gene Name
- Rad, Gem/Kir family member 1
- Gene Aliases
-
CG30127, CG44011, CG9811, CT27512, Dmel\CG44011, Dmel_CG44011, RGK1, rgk1
- Chromosome Location
-
Chr.2R: 19311148 - 19356525 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0264753
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_166341.3 |
NP_725875.3 |
5 - 6 |
1990 |
81 |
View Full Product Details