Assay Design
Gene Transcripts
- Gene Symbol
- Obp57d
- Entrez Gene ID
-
246671
- Gene Name
- Odorant-binding protein 57d
- Gene Aliases
-
57d, CG30150, DmelObp57d, Dmel\CG30150, Dmel\Obp57d, Dmel_CG30150, OBP57d, Obp-g, Obp57e, anon-AE003791.1
- Chromosome Location
-
Chr.2R: 20548368 - 20550117 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0043536
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_166397.3 |
NP_725973.1 |
4 - 5 |
894 |
130 |
- Gene Symbol
- Cpr57A
- Entrez Gene ID
-
37319
- Gene Name
- Cuticular protein 57A
- Gene Aliases
-
CG18066, Dmel\CG18066, Dmel_CG18066
- Chromosome Location
-
Chr.2R: 20548368 - 20550117 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034517
View Full Product Details