Assay Design



Gene Transcripts

Gene Symbol
Sara
Entrez Gene ID
44263
Gene Name
Smad anchor for receptor activation
Gene Aliases
CG15667, Dmel\CG15667, Dmel_CG15667, SARA, dSARA, sara
Chromosome Location
Chr.2R: 21481927 - 21487107 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026369
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079990.3 NP_524729.2 5 - 6 3774 78
GenBank mRNA AF239997.1 - 5 - 6 3751 78
AJ310804.1 - 5 - 6 3779 78
BT058000.1 - 5 - 6 3782 78

View Full Product Details