Assay Design
Gene Transcripts
- Gene Symbol
- PTP-ER
- Entrez Gene ID
-
37461
- Gene Name
- Protein tyrosine phosphatase-ERK/Enhancer of Ras1
- Gene Aliases
-
CG9856, Dmel\CG9856, Dmel_CG9856, ER2-5, ptp-er
- Chromosome Location
-
Chr.2R: 21619321 - 21624446 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0016641
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_166465.2 |
NP_726092.1 |
1 - 2 |
3117 |
73 |
GenBank mRNA |
AF146594.1 |
- |
1 - 2 |
3121 |
73 |
View Full Product Details