Assay Design



Gene Transcripts

Gene Symbol
Synj
Entrez Gene ID
37517
Gene Name
Synaptojanin
Gene Aliases
CG6562, Dmel\CG6562, Dmel_CG6562, IPP, synj
Chromosome Location
Chr.2R: 22080176 - 22085767 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034691
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259525.1 NP_001246454.1 2 - 3 1643 78
NM_130462.4 NP_569729.1 2 - 3 1594 78
NM_166505.2 NP_726155.1 2 - 3 1660 78
GenBank mRNA BT016135.1 - 2 - 3 1731 78

View Full Product Details