Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001103947.2 | NP_001097417.1 | 11 - 12 | 2138 | 61 |
NM_001274207.1 | NP_001261136.1 | 12 - 13 | 3786 | 61 | |
NM_001382123.1 | NP_001369111.1 | 12 - 13 | 3766 | 61 | |
NM_001382124.1 | NP_001369112.1 | 12 - 13 | 3766 | 61 | |
NM_166535.2 | NP_726208.1 | 12 - 13 | 3766 | 61 |