Assay Design
Gene Transcripts
- Gene Symbol
- Gr59a
- Entrez Gene ID
-
117485
- Gene Name
- Gustatory receptor 59a
- Gene Aliases
-
CG13537, CG30189, Dmel\CG30189, Dmel_CG30189, GR59C.1, GR59a
- Chromosome Location
-
Chr.2R: 23080561 - 23081725 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0045483
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_166585.2 |
NP_726287.2 |
1 - 2 |
943 |
84 |
View Full Product Details