Assay Design
Gene Transcripts
- Gene Symbol
- Or33b
- Entrez Gene ID
-
34602
- Gene Name
- Odorant receptor 33b
- Gene Aliases
-
33B.2, 33b, AN1, CG16961, DOR33B.2, DOR72, Dmel\CG16961, Dmel_CG16961, Or33B.2, Or72, dor72
- Chromosome Location
-
Chr.2L: 11936181 - 11937501 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026391
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078830.1 |
NP_523554.1 |
1 - 2 |
722 |
73 |
View Full Product Details