Assay Design
Gene Transcripts
- Gene Symbol
- Or33c
- Entrez Gene ID
-
34603
- Gene Name
- Odorant receptor 33c
- Gene Aliases
-
33B.3, 33c, AN2, CG5006, DOR33B.3, DOR71, Dmel\CG5006, Dmel_CG5006, Or33B.3, Or71, dor71
- Chromosome Location
-
Chr.2L: 11937969 - 11939202 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026390
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078831.1 |
NP_523555.1 |
1 - 2 |
732 |
78 |
View Full Product Details