Assay Design



Gene Transcripts

Gene Symbol
Tep1
Entrez Gene ID
34937
Gene Name
Thioester-containing protein 1
Gene Aliases
BG:DS00365.2, CG18096, Dmel\CG18096, Dmel_CG18096, TEP I, TEP1, TEP1mel, Tep 1, Tep I, Tep-1, TepI, dTEPI, tep1
Chromosome Location
Chr.2L: 15888640 - 15893811 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0041183
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078854.2 NP_523578.1 3 - 4 1597 82
GenBank mRNA AJ269538.1 - 3 - 4 1597 82

View Full Product Details